Läs mer
Mono[2-(carboxymethyl)hexyl] Phthalate, TRC
Produktkod. 30004320
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004320 1 mg
30012820 10 mg
30004516 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004320 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5255851MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82975-93-7
Molekylformel C16 H20 O6
Molekylvikt (g/mol) 308.33
Synonym DEHP Metabolite IV
IUPAC-namn 2-[2-(carboxymethyl)hexoxycarbonyl]benzoic acid
LEDER CCCCC(COC(=O)c1ccccc1C(=O)O)CC(=O)O
Specifikationer
Kemiskt namn eller material Mono[2-(carboxymethyl)hexyl] Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C16H20O6/c1-2-3-6-11(9-14(17)18)10-22-16(21)13-8-5-4-7-12(13)15(19)20/h4-5,7-8,11H,2-3,6,9-10H2,1H3,(H,17,18)(H,19,20)
Formel vikt 308.126
Procent renhet >95%