Läs mer
Mono-2-ethyl-5-carboxypentyl Terephthalate (MECPTP), TRC
Produktkod. 30133822
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133822 10 mg
30007692 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133822 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM52563010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1684398-42-2
Molekylformel C16 H20 O6
Molekylvikt (g/mol) 308.3264
Synonym Terephthalic Acid, Mono-2-ethyl-5-carboxypentyl Ester, 1,4-Benzenedicarboxylic acid, 1-(5-carboxy-2-ethylpentyl) ester, 1-(5-Carboxy-2-ethylpentyl) hydrogen 1,4-benzenedicarboxylate
IUPAC-namn 4-(5-carboxy-2-ethylpentoxy)carbonylbenzoic acid
LEDER CCC(CCCC(=O)O)COC(=O)c1ccc(cc1)C(=O)O
Specifikationer
Kemiskt namn eller material Mono-2-ethyl-5-carboxypentyl Terephthalate (MECPTP)
Kvantitet 10 mg
InChI formel InChI=1S/C16H20O6/c1-2-11(4-3-5-14(17)18)10-22-16(21)13-8-6-12(7-9-13)15(19)20/h6-9,11H,2-5,10H2,1H3,(H,17,18)(H,19,20)
Formel vikt 308.126