Läs mer
Mono(2-ethyl-5-hydroxyhexyl) Phthalate (Mixture of Diastereomers) (>90%), TRC
Produktkod. 17931235
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17931235 25 mg -
17920525 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17931235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM54251025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40321-99-1
Molekylformel C16 H22 O5
Molekylvikt (g/mol) 294.34
Synonym 1,2-Benzenedicarboxylic acid, mono(2-ethyl-5-hydroxyhexyl) ester (9CI), Mono-2-ethyl-5-hydroxyhexyl phthalate
IUPAC-namn 2-(2-ethyl-5-hydroxyhexoxy)carbonylbenzoic acid
LEDER CCC(CCC(C)O)COC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Mono(2-ethyl-5-hydroxyhexyl) Phthalate (Mixture of Diastereomers)
Kvantitet 25 mg
InChI formel InChI=1S/C16H22O5/c1-3-12(9-8-11(2)17)10-21-16(20)14-7-5-4-6-13(14)15(18)19/h4-7,11-12,17H,3,8-10H2,1-2H3,(H,18,19)
Formel vikt 294.1467