Läs mer
Mono-2-ethylhexyl adipate, TRC
Produktkod. 30466983
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30466983 1 g
1 options
Produktkod. 30466983 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM8648501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mono-2-ethylhexyl adipate
CAS 4337-65-9
Molekylformel C14H26O4
Kvantitet 1 g
Synonym Mono(2-Ethylhexyl)Adipate
InChI formel InChI=1S/C14H26O4/c1-3-5-8-12(4-2)11-18-14(17)10-7-6-9-13(15)16/h12H,3-11H2,1-2H3,(H,15,16)
LEDER O=C(O)CCCCC(=O)OCC(CC)CCCC
Molekylvikt (g/mol) 258.35