Learn More
Mono-2-ethylhexyl Phthalate Glucuronide, TRC
Product Code. 30133264
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133264 10 mg 10mg
30009996 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133264 Supplier Toronto Research Chemicals Supplier No. TRCM54250010MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59348-65-1
Molecular Formula C22H30O10
Molecular Weight (g/mol) 454.47
Synonym 1-(2-Ethylhexyl 1,2-Benzenedicarboxylate) beta-D-Glucopyranuronic Acid
IUPAC Name (2S,3S,5R,6S)-6-[2-(2-ethylhexoxycarbonyl)benzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CCCCC(CC)COC(=O)c1ccccc1C(=O)O[C@@H]2O[C@@H]([C@@H](O)C(O)[C@H]2O)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C22H30O10/c1-3-5-8-12(4-2)11-30-20(28)13-9-6-7-10-14(13)21(29)32-22-17(25)15(23)16(24)18(31-22)19(26)27/h6-7,9-10,12,15-18,22-25H,3-5,8,11H2,1-2H3,(H,26,27)/t12?,15?,16-,17+,18-,22-/m0/s1
Formula Weight 454.1839
Chemical Name or Material Mono-2-ethylhexyl Phthalate Glucuronide