Läs mer
Mono-2-hydroxybutyl Phthalate, TRC
Produktkod. 30482836
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482836 25 mg
30468927 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30482836 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM54751525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 856985-72-3
Molekylformel C12H14O5
Molekylvikt (g/mol) 238.24
Synonym 1,2-Benzenedicarboxylic Acid 1-(2-hydroxybutyl) Ester, 1,2-Butanediol Phthalate
IUPAC-namn 2-(2-hydroxybutoxycarbonyl)benzoic acid
LEDER O=C(C1=CC=CC=C1C(O)=O)OCC(O)CC
Specifikationer
Kemiskt namn eller material Mono-2-hydroxybutyl Phthalate
Kvantitet 25 mg
InChI formel InChI=1S/C12H14O5/c1-2-8(13)7-17-12(16)10-6-4-3-5-9(10)11(14)15/h3-6,8,13H,2,7H2,1H3,(H,14,15)
Formel vikt 238.0841