Läs mer
Mono(2-hydroxyisobutyl)phthalate, TRC
Produktkod. 30138334
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138334 100 mg
17984635 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138334 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM547525100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64339-39-5
Molekylformel C12H14O5
Molekylvikt (g/mol) 238.24
Synonym 1,2-Benzenedicarboxylic Acid 1-(2-Hydroxy-2-methylpropyl) Ester, 1,2-Benzenedicarboxylic Acid Mono(2-hydroxy-2-methylpropyl) Ester;
IUPAC-namn 2-(2-hydroxy-2-methylpropoxy)carbonylbenzoic acid
LEDER CC(=O)O[C@@H]1CC2[C@@](CC1)(C)[C@H]3[C@@H](CC=2)[C@@H]4[C@@](CC3)(C)C(c5cnccc5)=CC4
Specifikationer
Kemiskt namn eller material Mono(2-hydroxyisobutyl)phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H14O5/c1-12(2,16)7-17-11(15)9-6-4-3-5-8(9)10(13)14/h3-6,16H,7H2,1-2H3,(H,13,14)
Formel vikt 238.0841
Procent renhet >95%