Läs mer
Mono(3-hydroxybutyl)phthalate, TRC
Produktkod. 30010371
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010371 50 mg
30004253 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010371 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM54630050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57074-43-8
Molekylformel C13H16O4
Molekylvikt (g/mol) 236.26
Synonym 1,2-Benzenedicarboxylic Acid 1-(3-Hydroxybutyl)ester, 1,2-Benzenedicarboxylic Acid Mono(3-hydroxybutyl)ester, MHBP;
IUPAC-namn 2-(5-hydroxyhexanoyl)benzoic acid
LEDER CC(O)CCCC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Mono(3-hydroxybutyl)phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C13H16O4/c1-9(14)5-4-8-12(15)10-6-2-3-7-11(10)13(16)17/h2-3,6-7,9,14H,4-5,8H2,1H3,(H,16,17)
Formel vikt 236.1049
Procent renhet >95%