Läs mer
Mono(4-carboxybutyl) Phthalate-d4, TRC
Produktkod. 30471711
Change view
Click to view available options
Kvantitet:
2.5 mg
Unit Size:
2.5mg
Product Code. Kvantitet unitSize
30471711 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30471711 Supplier Toronto Research Chemicals Supplier No. TRCM5253222.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Mono(4-carboxybutyl) Phthalate-d4
CAS 1794737-39-5
Molekylformel C13H10D4O6
Kvantitet 2.5 mg
Synonym 1,2-(Benzene-d4)dicarboxylic Acid 1-(4-Carboxybutyl) Ester, MCBP-d4, 1,2-(Benzene-d4)dicarboxylic Acid Mono(4-carboxybutyl) Ester
InChI formel InChI=1S/C13H14O6/c14-11(15)7-3-4-8-19-13(18)10-6-2-1-5-9(10)12(16)17/h1-2,5-6H,3-4,7-8H2,(H,14,15)(H,16,17)/i1D,2D,5D,6D
LEDER O=C(OCCCCC(O)=O)C1=C([2H])C([2H])=C([2H])C([2H])=C1C(O)=O
Molekylvikt (g/mol) 270.27