Läs mer
Mono-(7-carboxy-2,7-dimethylheptyl) Phthalate, TRC
Produktkod. 30137807
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137807 10 mg -
30009942 5 mg -
30004039 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM52561510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1373125-93-9
Molekylformel C18H24O6
Molekylvikt (g/mol) 336.38
Synonym 1,2-Benzenedicarboxylic acid 1-(7-Carboxy-2-methyloctyl) Ester
IUPAC-namn 2-(7-carboxy-2-methyloctoxy)carbonylbenzoic acid
LEDER CC(CCCCC(C)C(=O)O)COC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Mono-(7-carboxy-2,7-dimethylheptyl) Phthalate
Kvantitet 10 mg
InChI formel InChI=1S/C18H24O6/c1-12(7-3-4-8-13(2)16(19)20)11-24-18(23)15-10-6-5-9-14(15)17(21)22/h5-6,9-10,12-13H,3-4,7-8,11H2,1-2H3,(H,19,20)(H,21,22)
Formel vikt 336.1573