Läs mer
Mono-7-carboxyheptyl Phthalate, TRC
Produktkod. 30005655
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005655 250 mg
30009138 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005655 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM525580250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 856869-57-3
Molekylformel C16H20O6
Molekylvikt (g/mol) 308.33
Synonym Mono-7-carboxyheptyl phthalate, 2-{[(7-carboxyheptyl)oxy]carbonyl}benzoic acid, 1,2-benzenedicarboxylic acid, mono-7-carboxyheptyl ester
IUPAC-namn 2-(7-carboxyheptoxycarbonyl)benzoic acid
LEDER C1=CC=C(C(=C1)C(=O)O)C(=O)OCCCCCCCC(=O)O
Specifikationer
Kemiskt namn eller material Mono(7-carboxyheptyl) phthalate
Kvantitet 250 mg
InChI formel InChI=1S/C16H20O6/c17-14(18)10-4-2-1-3-7-11-22-16(21)13-9-6-5-8-12(13)15(19)20/h5-6,8-9H,1-4,7,10-11H2,(H,17,18)(H,19,20)
Formel vikt 308.126