Läs mer
Mono-carboxy-isooctyl Phthalate, TRC
Produktkod. 30006870
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006870 10 mg
30006854 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006870 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM52559010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 898544-09-7
Molekylformel C17H22O6
Molekylvikt (g/mol) 322.35
Synonym 2-(((8-Carboxyoctan-2-yl)oxy)carbonyl)benzoic Acid
IUPAC-namn 2-(7-carboxy-1-methyl-heptoxy)carbonylbenzoic acid
LEDER CC(CCCCCCC(=O)O)OC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Mono-carboxy-isooctyl Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C17H22O6/c1-12(8-4-2-3-5-11-15(18)19)23-17(22)14-10-7-6-9-13(14)16(20)21/h6-7,9-10,12H,2-5,8,11H2,1H3,(H,18,19)(H,20,21)
Formel vikt 322.1416
Procent renhet >95%