Läs mer
Mono-POC Ethyl Tenofovir (Mixture of Diastereomers), TRC
Produktkod. 30481473
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30481473 1 mg
1 options
Produktkod. 30481473 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5669851MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mono-POC Ethyl Tenofovir (Mixture of Diastereomers)
CAS 1796539-92-8
Molekylformel C16H26N5O7P
Kvantitet 1 mg
Synonym (8R)-9-(6-Amino-9H-purin-9-yl)-5-ethoxy-8-methyl-2,4,7-trioxa-5-phosphanonanoic Acid Methyl 1-Methylethyl Ester 5-Oxide, Tenofovir Ethyl Monoisoproxil, [Isopropylcarbonyloxymethyl-(Ethoxy)-[(R)-9-(2-phosphonomethoxy)]propyl Adenine
InChI formel InChI=1S/C16H26N5O7P/c1-5-26-29(23,27-9-24-16(22)28-11(2)3)10-25-12(4)6-21-8-20-13-14(17)18-7-19-15(13)21/h7-8,11-12H,5-6,9-10H2,1-4H3,(H2,17,18,19)/t12-,29?/m1/s1
LEDER CC(C)OC(OCOP(CO[C@H](C)CN1C2=NC=NC(N)=C2N=C1)(OCC)=O)=O
IUPAC-namn ((((((R)-1-(6-amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)(ethoxy)phosphoryl)oxy)methyl isopropyl carbonate
Molekylvikt (g/mol) 431.38
Formel vikt 431.16