Läs mer
Mono-POC Tenofovir 6-Isopropyl Carbamate (Mixture of Diastereomers), TRC
Produktkod. 30465268
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30465268 1 mg
1 options
Produktkod. 30465268 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5670101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mono-POC Tenofovir 6-Isopropyl Carbamate (mixture of diastereomers)
CAS 1244022-56-7
Molekylformel C18 H28 N5 O9 P
Kvantitet 1 mg
Synonym (8R)-5-hydroxy-8-methyl-9-[6-[[(1-methylethoxy)carbonyl]amino]-9H-purin-9-yl]-2,4,7-trioxa-5-phosphanonanoic acid 1-methylethyl ester 5-oxide, [(1R)-2-[6-(1-(Methyl)ethoxycarbonylamino)-9H-purin-9-yl]-1-(methyl)ethoxy]methyl-(1-(methyl)ethoxycarbonyloxymethoxy)phosphinic Acid, [(2R)-3-[6-(Isopropyloxycarbonylamino)-9H-purin-9-yl]prop-2-yloxy]methyl(isopropyloxycarbonyloxymethoxy)phosphinic Acid, [(2R)-1-[6-(Isopropyloxycarbonylamino)-9H-purin-9-yl]propan-2-yloxy]methyl(isopropyloxycarbonyloxymethoxy)phosphinic Acid, [[[(2R)-1-[6-(Isopropyloxycarbonylamino)-9H-purin-9-yl]propan-2-yloxy]methyl](hydroxy)phosphoryloxy]methyl Isopropyl Carbonate
InChI formel InChI=1S/C18H28N5O9P/c1-11(2)31-17(24)22-15-14-16(20-7-19-15)23(8-21-14)6-13(5)29-10-33(26,27)30-9-28-18(25)32-12(3)4/h7-8,11-13H,6,9-10H2,1-5H3,(H,26,27)(H,19,20,22,24)/t13-/m1/s1
LEDER CC(C)OC(=O)Nc1ncnc2c1ncn2C[C@@H](C)OCP(=O)(O)OCOC(=O)OC(C)C
IUPAC-namn [(2R)-1-[6-(propan-2-yloxycarbonylamino)purin-9-yl]propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphinic acid
Molekylvikt (g/mol) 489.42
Formel vikt 489.1625