Läs mer
Mono-POC Tenofovir, TRC
Produktkod. 30011918
Change view
Kvantitet:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011918 100 mg
30003953 10 mg
17993845 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM567000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 211364-69-1
Molekylformel C14 H22 N5 O7 P
Molekylvikt (g/mol) 403.33
Synonym 2,4,7-Trioxa-5-phosphanonanoic acid, 9-(6-amino-9H-purin-9-yl)-5-hydroxy-8-methyl-, 1-methylethyl ester, 5-oxide, (8R)-, Tenofovir monoisoproxil
IUPAC-namn [(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphinic acid
LEDER CC(C)OC(=O)OCOP(=O)(O)CO[C@H](C)Cn1cnc2c(N)ncnc12
Specifikationer
Kemiskt namn eller material Mono-POC Tenofovir (Mixture of Diastereomers)
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H22N5O7P/c1-9(2)26-14(20)23-7-25-27(21,22)8-24-10(3)4-19-6-18-11-12(15)16-5-17-13(11)19/h5-6,9-10H,4,7-8H2,1-3H3,(H,21,22)(H2,15,16,17)/t10-/m1/s1
Formel vikt 403.1257
Procent renhet >95%