Läs mer
Mono-sec-butyl Phthalate, TRC
Produktkod. 30472132
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472132 2.5 g
30466243 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30472132 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5251102.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53623-59-9
Molekylformel C12H14O4
Molekylvikt (g/mol) 222.24
Synonym 1,2-Benzenedicarboxylic Acid 1-(1-methylpropyl) Ester, 1,2-Benzenedicarboxylic Acid Mono(1-methylpropyl) Ester, Phthalic Acid sec-Butyl Ester, NSC 29059
IUPAC-namn 2-butan-2-yloxycarbonylbenzoic acid
LEDER CCC(C)OC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Mono-sec-butyl Phthalate
Kvantitet 2.5 g
InChI formel InChI=1S/C12H14O4/c1-3-8(2)16-12(15)10-7-5-4-6-9(10)11(13)14/h4-8H,3H2,1-2H3,(H,13,14)
Formel vikt 222.0892