Läs mer
Monobehenin, MedChemExpress
Produktkod. 16451906
Encompass_Preferred
Change view
Kvantitet:
100 mg
500 mg
Förpackningsstorlek:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16451906 500 mg -
16407796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16451906 Leverantör Medchem Express Leverantörsnummer HY20349
MedChemExpress Monobehenin, an bacterial biofilm formation inhibitor, has strong inhibitory activity toward bacterial biofilm formation of S. mutans, X. oryzae, and Y. enterocolitica in a strain specific manner.
Monobehenin, an bacterial biofilm formation inhibitor, has strong inhibitory activity toward bacterial biofilm formation of S. mutans, X. oryzae, and Y. enterocolitica in a strain specific manner.
Specifikationer
Kemiskt namn eller material Monobehenin
Fysisk form Solid
CAS 30233-64-8
Färg White
Kvantitet 500 mg
Anteckningar om renhetsgrad Research
Molekylformel C25H52O5
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation DMSO : ≥ 1 mg/mL (2.31 mM) ∣H2O : 0.1 mg/mL (0.23 mM; Need ultrasonic)
LEDER CCCCCCCCCCCCCCCCCCCCCC(O)=O.OCC(O)CO
Molekylvikt (g/mol) 432.68
Formel vikt 432.68