Läs mer
Monobenzyl Adipate, TRC
Produktkod. 30480826
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30480826 10 g
1 options
Produktkod. 30480826 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM52490510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monobenzyl Adipate
CAS 40542-90-3
Molekylformel C13 H16 O4
Kvantitet 10 g
Synonym Adipic Acid Benzyl Ester, Hexanedioic Acid Mono(phenylmethyl) Ester, 5-(Benzyloxycarbonyl)pentanoic Acid, Adipic Acid Monobenzyl Ester, Hexanedioic Acid Monobenzyl Ester
InChI formel InChI=1S/C13H16O4/c14-12(15)8-4-5-9-13(16)17-10-11-6-2-1-3-7-11/h1-3,6-7H,4-5,8-10H2,(H,14,15)
LEDER OC(=O)CCCCC(=O)OCc1ccccc1
IUPAC-namn 6-oxo-6-phenylmethoxyhexanoic acid
Molekylvikt (g/mol) 236.26
Formel vikt 236.1049