Läs mer
Monobenzyl Malonate, TRC
Produktkod. 30483670
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30483670 10 g
1 options
Produktkod. 30483670 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM52488010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monobenzyl Malonate
CAS 40204-26-0
Molekylformel C10H10O4
Kvantitet 10 g
Synonym Propanedioic Acid 1-(Phenylmethyl) Ester, Propanedioic Acid Mono(phenylmethyl) Ester, (Benzyloxycarbonyl)acetic acid, 2-(Benzyloxycarbonyl)acetic acid, 3-(Benzyloxy)-3-oxopropanoic acid, Benzyl hemimalonate, Benzyl hydrogen malonate, Malonic acid monobenzyl ester
InChI formel InChI=1S/C10H10O4/c11-9(12)6-10(13)14-7-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)
LEDER OC(=O)CC(=O)OCc1ccccc1
IUPAC-namn 3-oxo-3-phenylmethoxypropanoic acid
Molekylvikt (g/mol) 194.18
Formel vikt 194.0579