Läs mer
Monobenzyl Terephthalate, TRC
Produktkod. 30467633
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30467633 100 mg
30477631 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30467633 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM524930100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18520-63-3
Molekylformel C15H12O4
Molekylvikt (g/mol) 256.25
Synonym 1,4-Benzenedicarboxylic Acid Mono(phenylmethyl) Ester, Terephthalic Acid Benzyl Ester, Terephthalic Acid Monobenzyl Ester, Monobenzyl 1,4-Benzenedicarboxylate, Monobenzyl Terephthalate, p-(Benzyloxycarbonyl)benzoic Acid
LEDER COc1cc2c(cc1)c3c(c(C)ncc3)n2
Specifikationer
Kemiskt namn eller material Monobenzyl Terephthalate
Kvantitet 100 mg
InChI formel InChI=1S/C15H12O4/c16-14(17)12-6-8-13(9-7-12)15(18)19-10-11-4-2-1-3-5-11/h1-9H,10H2,(H,16,17)