Läs mer
Monobutyl Phosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Monobutyl Phosphate |
| CAS | 1623-15-0 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C4H11O4P |
| Kvantitet | 50 mg |
| Synonym | Phosphoric Acid Monobutyl Ester, Butyl Diacid Phosphate, Butyl Dibasic Acid Phosphate, Butyl Dihydrogen Phosphate, Dihydrogen Monobutyl Phosphate, JAMP 4P, MP 4, Monobutyl Dihydrogen Phosphate, Monobutyl Phosphate, NSC 41911 |
| InChI formel | InChI=1S/C4H11O4P/c1-2-3-4-8-9(5,6)7/h2-4H2,1H3,(H2,5,6,7) |
| LEDER | CCCCOP(=O)(O)O |
| IUPAC-namn | butyl dihydrogen phosphate |
| Molekylvikt (g/mol) | 154.1 |
| Visa mer |