Läs mer
Monocarboxy Isononyl Phthalate, TRC
Produktkod. 17930525
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17930525 10 mg
17941645 50 mg
17912415 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17930525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM54792010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2042194-73-8
Molekylformel C18H24O6
Molekylvikt (g/mol) 336.38
Synonym 2-(((9-Carboxynonyl)oxy)carbonyl)benzoic Acid
IUPAC-namn 2-(9-carboxynonoxycarbonyl)benzoic acid
LEDER OC(=O)CCCCCCCCCOC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Monocarboxy Isononyl Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C18H24O6/c19-16(20)12-6-4-2-1-3-5-9-13-24-18(23)15-11-8-7-10-14(15)17(21)22/h7-8,10-11H,1-6,9,12-13H2,(H,19,20)(H,21,22)
Formel vikt 336.1573
Procent renhet >95%