Läs mer
Monodecarboxy Piperacilloic Acid (Mixture of Diastereomers), TRC
Produktkod. 30464664
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30464664 25 mg
1 options
Produktkod. 30464664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM52670025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monodecarboxy Piperacilloic Acid (Mixture of Diastereomers)
CAS 64817-23-8
Molekylformel C22 H29 N5 O6 S
Kvantitet 25 mg
Synonym α-(4-Ethyl-2,3-dioxo-1-piperazinecarboxamido)benzylpenilloic Acid, [2R-[2α(R*),4β]]-2-[[[[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]methyl]-5,5-dimethyl-4-thiazolidinecarboxylic Acid, Piperacillin EP Impurity C
InChI formel InChI=1S/C22H29N5O6S/c1-4-26-10-11-27(19(30)18(26)29)21(33)25-15(13-8-6-5-7-9-13)17(28)23-12-14-24-16(20(31)32)22(2,3)34-14/h5-9,14-16,24H,4,10-12H2,1-3H3,(H,23,28)(H,25,33)(H,31,32)/t14-,15-,16+/m1/s1
LEDER CCN1CCN(C(=O)N[C@@H](C(=O)NC[C@@H]2N[C@@H](C(=O)O)C(C)(C)S2)c3ccccc3)C(=O)C1=O
IUPAC-namn (2R,4S)-2-[[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]methyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molekylvikt (g/mol) 491.561
Formel vikt 491.184