Läs mer
Monodocosahexaenoin, TRC
Produktkod. 30480142
Change view
Click to view available options
Kvantitet:
10 mg
Product Code. Kvantitet
30480142 10 mg
1 options
Product Code. 30480142 Supplier Toronto Research Chemicals Supplier No. TRCM53955310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Monodocosahexaenoin
CAS 140670-40-2
Molekylformel C25H38O4
Kvantitet 10 mg
Synonym (4Z,7Z,10Z,13Z,16Z,19Z)-2,3-Dihydroxypropyl docosa-4,7,10,13,16,19-hexaenoate, Docosahexaenoylglycerol
InChI formel InChI=1S/C25H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-25(28)29-23-24(27)22-26/h3-4,6-7,9-10,12-13,15-16,18-19,24,26-27H,2,5,8,11,14,17,20-23H2,1H3/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-
LEDER O=C(OCC(O)CO)CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC
IUPAC-namn 2,3-dihydroxypropyl (4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoate
Molekylvikt (g/mol) 402.57
Formel vikt 402.28