Läs mer
Monoethyl Phthalate O-Beta-D-Glucuronide, TRC
Produktkod. 30465459
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30465459 5 mg
1 options
Produktkod. 30465459 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5425855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monoethyl Phthalate O-beta-D-Glucuronide
CAS 671215-25-1
Molekylformel C16H18O10
Kvantitet 5 mg
Synonym 1-(Ethyl 1,2-Benzenedicarboxylate) β-D-Glucopyranuronic Acid
InChI formel InChI=1S/C16H18O10/c1-2-24-14(22)7-5-3-4-6-8(7)15(23)26-16-11(19)9(17)10(18)12(25-16)13(20)21/h3-6,9-12,16-19H,2H2,1H3,(H,20,21)/t9-,10-,11+,12-,16-/m0/s1/i3D,4D,5D,6D
LEDER CCOC(=O)C1=CC=CC=C1C(=O)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C(=O)O)O)O)O
Molekylvikt (g/mol) 370.31