Läs mer
Monoethyl Phthalate, TRC
Produktkod. 30009950
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009950 50 mg
30007401 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM54258050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2306-33-4
Molekylformel C10 H10 O4
Molekylvikt (g/mol) 194.18
Synonym 1,2-Benzenedicarboxylic acid, 1-ethyl ester, 1,2-Benzenedicarboxylic acid, monoethyl ester (9CI), Phthalic acid, ethyl ester (6CI,7CI), Phthalic acid, monoethyl ester (8CI), Ethyl 2-carboxybenzoate, Monoethyl phthalate
IUPAC-namn 2-ethoxycarbonylbenzoic acid
LEDER CCOC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Monoethyl Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C10H10O4/c1-2-14-10(13)8-6-4-3-5-7(8)9(11)12/h3-6H,2H2,1H3,(H,11,12)
Formel vikt 194.0579
Procent renhet >95%