Läs mer
Monohydrazide Adipic Acid, TRC
Produktkod. 30479662
Change view
Kvantitet:
100 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479662 100 mg
30467892 500 mg
30471831 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479662 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM547925100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6292-67-7
Molekylformel C6H12N2O3
Molekylvikt (g/mol) 160.17
Synonym 1-Hydrazidehexanedioic Acid, Monohydrazide Hexanedioic Acid, NSC 9926
IUPAC-namn 6-hydrazinyl-6-oxohexanoic acid
LEDER NNC(=O)CCCCC(=O)O
Specifikationer
Kemiskt namn eller material Monohydrazide Adipic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C6H12N2O3/c7-8-5(9)3-1-2-4-6(10)11/h1-4,7H2,(H,8,9)(H,10,11)
Formel vikt 160.0848