Läs mer
Monohydroxyisononyl Phthalate, TRC
Produktkod. 30134581
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134581 100 mg
30007660 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134581 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM547540100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2741633-20-3
Molekylformel C17H24O5
Molekylvikt (g/mol) 308.37
IUPAC-namn 2-(((9-hydroxynonan-2-yl)oxy)carbonyl)benzoic acid
LEDER CC(CCCCCCCO)OC(C1=CC=CC=C1C(O)=O)=O
Specifikationer
Kemiskt namn eller material Monohydroxyisononyl Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C17H24O5/c1-13(9-5-3-2-4-8-12-18)22-17(21)15-11-7-6-10-14(15)16(19)20/h6-7,10-11,13,18H,2-5,8-9,12H2,1H3,(H,19,20)
Formel vikt 308.16
Procent renhet >95%