Läs mer
Monolaurin, TRC
Produktkod. 30472410
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30472410 10 g
1 options
Produktkod. 30472410 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM54900010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monolaurin
CAS 142-18-7
Molekylformel C15H30O4
Kvantitet 10 g
Synonym 1-Mono-laurin, (±)-2,3-Dihydroxypropyl dodecanoate, (±)-Glyceryl 1-monododecanoate, 1-Glyceryl laurate, 1-Monododecanoylglycerol, 1-Monolaurin, 1-Monolauroyl-rac-glycerol, 3-Dodecanoyloxy-1,2-propanediol, Dodecanoic acid alpha-monoglyceride, Glycerin 1-monolaurate, Glycerol 1-laurate, Glycerol 1-monododecanoate, Glycerol 1-monolaurate, Glycerol alpha-monolaurate, Glyceryl laurate, Glyceryl monododecanoate, Glyceryl monolaurate, Lauric acid 1-monoglyceride, Lauric acid alpha-monoglyceride, Lauricidin, Luaricidin, NSC 698570, alpha-Monolaurin
InChI formel InChI=1S/C15H30O4/c1-2-3-4-5-6-7-8-9-10-11-15(18)19-13-14(17)12-16/h14,16-17H,2-13H2,1H3
LEDER CCCCCCCCCCCC(=O)OCC(O)CO
IUPAC-namn 2,3-dihydroxypropyl dodecanoate
Molekylvikt (g/mol) 274.4
Formel vikt 274.2144