Läs mer
Monomethyl Glutarate-1,5-13C2, TRC
Produktkod. 30464970
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30464970 50 mg
1 options
Produktkod. 30464970 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM56650250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monomethyl Glutarate-1,5-13C2
CAS 1189708-76-6
Molekylformel C413C2H10O4
Kvantitet 50 mg
Synonym Pentanedioic Acid-13C2 1-Methyl Ester, Glutaric Acid-13C2 Monomethyl Ester, 4-(Methoxycarbonyl)butyric Acid-13C2, 4-Carboxybutanoic Acid-13C2 Methyl Ester, 5-Methoxy-5-oxopentanoic Acid-13C2, Methyl Glutarate-13C2, NSC 93807-13C2
InChI formel InChI=1S/C6H10O4/c1-10-6(9)4-2-3-5(7)8/h2-4H2,1H3,(H,7,8)/i5+1,6+1
LEDER CO[13C](=O)CCC[13C](=O)O
IUPAC-namn 5-methoxy-5-oxo(1,5-^{13}C_{2})pentanoic acid
Molekylvikt (g/mol) 148.13
Formel vikt 148.0646