Läs mer
Monomethyl Succinate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Monomethyl Succinate |
| CAS | 3878-55-5 |
| Molekylformel | C5H8O4 |
| Kvantitet | 5 g |
| Synonym | 3-Carbomethoxypropanoic acid, 4-Methoxy-4-oxobutanoic Acid, Methyl Hydrogen Succinate, Methyl Monosuccinate, Monomethyl Butanedioate, Monomethyl Succinate NSC 511, 3-(Methoxycarbonyl)propanoic Acid, 3-(Methoxycarbonyl)propionic Acid, Monomethyl Succinic Acid Ester, Methyl Succinic Acid Ester, Monomethyl Butanedioic Acid Ester |
| InChI formel | InChI=1S/C5H8O4/c1-9-5(8)3-2-4(6)7/h2-3H2,1H3,(H,6,7) |
| LEDER | COC(=O)CCC(=O)O |
| IUPAC-namn | 4-methoxy-4-oxobutanoic acid |
| Molekylvikt (g/mol) | 132.11 |
| Formel vikt | 132.0423 |