Läs mer
α-Monomyristin, TRC
Produktkod. 30476093
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30476093 250 mg
1 options
Produktkod. 30476093 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM566560250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Alpha-Monomyristin
CAS 589-68-4
Molekylformel C17 H34 O4
Kvantitet 250 mg
Synonym Tetradecanoic Acid 2,3-Dihydroxypropyl Ester, ()-2,3-Dihydroxypropyl Tetradecanoate, 1-Monomyristin, 1-Monomyristoyl-Rac-Glycerol, 1-Monotetradecanoylglycerol, Glycerol 1-Monotetradecanoate, Glycerol 1-Myristate, Glyceryl Myristate, Myristic Acid 1-Monoglyceride, Nsc 404225
InChI formel InChI=1S/C17H34O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-17(20)21-15-16(19)14-18/h16,18-19H,2-15H2,1H3
LEDER Cccccccccccccc(=O)Occ(O)Co
IUPAC-namn 2,3-dihydroxypropyl tetradecanoate
Molekylvikt (g/mol) 302.45
Formel vikt 302.2457