Läs mer
Monosodium Acid Methane Arsonate Sesquihydrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methylarsonic acid sodium |
| CAS | 2163-80-6 |
| Molekylformel | C H4 As O3 . Na |
| Kvantitet | 25 mg |
| Synonym | Methanearsonic acid, monosodium salt (8CI), Ansar 170, Ansar 170L, Ansar 529, Bueno 6, Daconate, Gepiron, MSMA, Monosodium acid methanearsonate, Monosodium acid metharsonate, Monosodium methanearsonate, Monosodium methylarsonate, Sodium acid methanearsonate, Weed 108 |
| InChI formel | InChI=1S/CH5AsO3.Na/c1-2(3,4)5;/h1H3,(H2,3,4,5);/q;+1/p-1 |
| LEDER | [Na+].C[As](=O)(O)[O-] |
| IUPAC-namn | sodium;hydroxy(methyl)arsinate |
| Molekylvikt (g/mol) | 161.952 |
| Formel vikt | 161.9274 |