Läs mer
Monosulfuron Ester, TRC
Produktkod. 30482688
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30482688 500 mg
30478826 250 mg
30477079 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30482688 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM175070500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 175076-90-1
Molekylformel C14 H14 N4 O5 S
Molekylvikt (g/mol) 350.3498
Synonym Monosulfuron-ester, Benzoic acid, 2-[[[[(4-methyl-2-pyrimidinyl)amino]carbonyl]amino]sulfonyl]-, methyl ester, Monosulfuron ester, Monosulfuron-methyl, NK 94827
IUPAC-namn methyl 2-[(4-methylpyrimidin-2-yl)carbamoylsulfamoyl]benzoate
LEDER COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nccc(C)n2
Specifikationer
Kemiskt namn eller material Monosulfuron-ester
Kvantitet 500 mg
InChI formel InChI=1S/C14H14N4O5S/c1-9-7-8-15-13(16-9)17-14(20)18-24(21,22)11-6-4-3-5-10(11)12(19)23-2/h3-8H,1-2H3,(H2,15,16,17,18,20)
Formel vikt 350.0685