Läs mer
Monosulfuron, TRC
Produktkod. 30479062
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479062 250 mg
30478715 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479062 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM567185250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monosulfuron
CAS 155860-63-2
Molekylformel C12 H11 N5 O5 S
Kvantitet 250 mg
Synonym Benzenesulfonamide, N-[[(4-methyl-2-pyrimidinyl)amino]carbonyl]-2-nitro-, Monosulfuron
InChI formel InChI=1S/C12H11N5O5S/c1-8-6-7-13-11(14-8)15-12(18)16-23(21,22)10-5-3-2-4-9(10)17(19)20/h2-7H,1H3,(H2,13,14,15,16,18)
LEDER Cc1ccnc(NC(=O)NS(=O)(=O)c2ccccc2[N+](=O)[O-])n1
IUPAC-namn 1-(4-methylpyrimidin-2-yl)-3-(2-nitrophenyl)sulfonylurea
Molekylvikt (g/mol) 337.31
Formel vikt 337.0481