Läs mer
Monosultap, TRC
Produktkod. 30469543
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30469543 100 mg
1 options
Produktkod. 30469543 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM567180100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Monosultap
CAS 29547-00-0
Molekylformel C5 H12 N O6 S4 . Na
Kvantitet 100 mg
Synonym Thiosulfuric acid (H2S2O3), S,S'-[2-(dimethylamino)-1,3-propanediyl] ester, monosodium salt (9CI), Thiosulfuric acid, S,S'-[2-(dimethylamino)trimethylene] ester, monosodium salt (8CI), 1,3-Propanedithiol, 2-(dimethylamino)-, bis(hydrogen sulfate) (ester) monosodium salt (8CI), Monomehypo, Monosultap, SCD, Sha Chong Dan, Shachongdan
InChI formel InChI=1S/C5H13NO6S4.Na/c1-6(2)5(3-13-15(7,8)9)4-14-16(10,11)12;/h5H,3-4H2,1-2H3,(H,7,8,9)(H,10,11,12);/q;+1/p-1
LEDER [Na+].CN(C)C(CSS(=O)(=O)O)CSS(=O)(=O)[O-]
IUPAC-namn sodium;2-(dimethylamino)-1-sulfonatosulfanyl-3-sulfosulfanylpropane
Molekylvikt (g/mol) 333.4
Formel vikt 332.9445