Läs mer
Montelukast 1,2-Diol (Mixture of Diastereomers), TRC
Produktkod. 30476849
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30476849 1 mg
1 options
Produktkod. 30476849 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM5680151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Montelukast 1,2-Diol(Mixture of Diastereomers)
CAS 186352-97-6
Molekylformel C35 H36 Cl N O4 S
Kvantitet 1 mg
Synonym Cyclopropaneacetic acid, 1-[[[(1R)-1-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1,2-dihydroxy-1-methylethyl)phenyl]propyl]thio]methyl]- (9CI, ACI), 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1,2-dihydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic acid (ACI), Cyclopropaneacetic acid, 1-[[[1-[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1,2-dihydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, [1R-[1R*(E)]]-[partial]- (ZCI)
InChI formel InChI=1S/C35H36ClNO4S/c1-23(38)34(41)30-8-3-2-6-25(30)12-16-32(42-22-35(17-18-35)21-33(39)40)27-7-4-5-24(19-27)9-14-29-15-11-26-10-13-28(36)20-31(26)37-29/h2-11,13-15,19-20,23,32,34,38,41H,12,16-18,21-22H2,1H3,(H,39,40)/b14-9+/t23?,32-,34?/m1/s1
LEDER CC(O)C(O)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3
IUPAC-namn 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1,2-dihydroxypropyl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetic acid
Molekylvikt (g/mol) 602.18
Formel vikt 601.2054