Läs mer
Montelukast Cyclized Ether impurity, TRC
Produktkod. 30472527
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30472527 50 mg
1 options
Produktkod. 30472527 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM56805050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Montelukast Cyclizate
CAS 168214-67-3
Molekylformel C29 H26 Cl N O
Kvantitet 50 mg
Synonym Quinoline, 7-chloro-2-[2-[3-(1,3,4,5-tetrahydro-1,1-dimethyl-2-benzoxepin-3-yl)phenyl]ethenyl]-, 7-Chloro-2-{2-[3-(1,1-dimethyl-1,3,4,5-tetrahydrobenzo[c]-oxepin-3-yl)phenyl]vinyl}quinoline, Montelukast Cyclizate Ether Impurity, Montelukast Cyclizate
InChI formel InChI=1S/C29H26ClNO/c1-29(2)26-9-4-3-7-21(26)13-17-28(32-29)23-8-5-6-20(18-23)10-15-25-16-12-22-11-14-24(30)19-27(22)31-25/h3-12,14-16,18-19,28H,13,17H2,1-2H3
LEDER CC1(C)OC(CCc2ccccc12)c3cccc(C=Cc4ccc5ccc(Cl)cc5n4)c3
IUPAC-namn 7-chloro-2-[2-[3-(1,1-dimethyl-4,5-dihydro-3H-2-benzoxepin-3-yl)phenyl]ethenyl]quinoline
Molekylvikt (g/mol) 439.98
Formel vikt 439.1703