Läs mer
Montelukast Methyl Ketone, TRC
Produktkod. 30012334
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012334 50 mg
17994965 25 mg
17993365 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012334 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM56804050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 937275-23-5
Molekylformel C34 H32 Cl N O3 S
Molekylvikt (g/mol) 570.14
Synonym 1-[[[(1R)-3-(2-Acetylphenyl)-1-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]propyl]thio]methyl]-cyclopropaneacetic Acid, Montekulast Sodium Imp. F (EP), Montelukast Methyl Ketone, Mok-3 Keto
IUPAC-namn 2-[1-[[(1R)-3-(2-acetylphenyl)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]propyl]sulfanylmethyl]cyclopropyl]acetic acid
LEDER CC(=O)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3
Specifikationer
Kemiskt namn eller material Montelukast Methyl Ketone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C34H32ClNO3S/c1-23(37)30-8-3-2-6-25(30)12-16-32(40-22-34(17-18-34)21-33(38)39)27-7-4-5-24(19-27)9-14-29-15-11-26-10-13-28(35)20-31(26)36-29/h2-11,13-15,19-20,32H,12,16-18,21-22H2,1H3,(H,38,39)/b14-9+/t32-/m1/s1
Formel vikt 569.1791
Procent renhet >95%