missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morpholin-3-one, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 106.0°C to 108.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 5 g |
Formula Weight | 101.11 |
Percent Purity | 98% |
Chemical Name or Material | Morpholin-3-one |
Safety and Handling
Product Identifier
- Morpholin-3-one
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'