Läs mer
Morphothion, TRC
Produktkod. 30470848
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470848 500 mg
30470480 1 g
30467808 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30470848 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM414630500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 144-41-2
Molekylformel C8 H16 N O4 P S2
Molekylvikt (g/mol) 285.32
Synonym Phosphorodithioic acid, O,O-dimethyl S-(morpholinocarbonylmethyl) ester (6CI), Phosphorodithioic acid, O,O-dimethyl ester, S-ester with 4-(mercaptoacetyl)morpholine (7CI,8CI), Morpholine, 4-(mercaptoacetyl)-, S-ester with O,O-dimethyl phosphorodithioate (8CI), 4-(Mercaptoacetyl)morpholine O,O-dimethyl phosphorodithioate, Ekatin F, Ekatin M, Morphothion, Morphotox, O,O-Dimethyl S-(morpholinocarbonylmethyl) phosphorodithioate
IUPAC-namn 2-dimethoxyphosphinothioylsulfanyl-1-morpholin-4-ylethanone
LEDER COP(=S)(OC)SCC(=O)N1CCOCC1
Specifikationer
Kemiskt namn eller material Morphothion
Kvantitet 500 mg
InChI formel InChI=1S/C8H16NO4PS2/c1-11-14(15,12-2)16-7-8(10)9-3-5-13-6-4-9/h3-7H2,1-2H3
Formel vikt 285.0258