Läs mer
Morusin, TRC
Produktkod. 30136325
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136325 100 mg
30003514 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136325 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM728000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 62596-29-6
Molekylformel C25 H24 O6
Molekylvikt (g/mol) 420.45
Synonym 4H,8H-Benzo[1,2-b:3,4-b']dipyran-4-one, 2-(2,4-dihydroxyphenyl)-5-hydroxy-8,8-dimethyl-3-(3-methyl-2-buten-1-yl)-, 4H,8H-Benzo[1,2-b:3,4-b']dipyran-4-one, 2-(2,4-dihydroxyphenyl)-5-hydroxy-8,8-dimethyl-3-(3-methyl-2-butenyl)- (9CI), 2-(2,4-Dihydroxyphenyl)-5-hydroxy-8,8-dimethyl-3-(3-methyl-2-buten-1-yl)-4H,8H-benzo[1,2-b:3,4-b']dipyran-4-one, Morusin, Mulberrochromene, NSC 649220
IUPAC-namn 2-(2,4-dihydroxyphenyl)-5-hydroxy-8,8-dimethyl-3-(3-methylbut-2-enyl)pyrano[2,3-h]chromen-4-one
LEDER CC(=CCC1=C(Oc2c3C=CC(C)(C)Oc3cc(O)c2C1=O)c4ccc(O)cc4O)C
Specifikationer
Kemiskt namn eller material Morusin
Kvantitet 100 mg
InChI formel InChI=1S/C25H24O6/c1-13(2)5-7-17-22(29)21-19(28)12-20-16(9-10-25(3,4)31-20)24(21)30-23(17)15-8-6-14(26)11-18(15)27/h5-6,8-12,26-28H,7H2,1-4H3
Formel vikt 420.1573