Läs mer
Mosapride N-Oxide, TRC
Produktkod. 30478983
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30478983 10 mg -
30472964 5 mg -
30470985 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30478983 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM73101510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1161443-73-7
Molekylformel C21 H25 Cl F N3 O4
Molekylvikt (g/mol) 437.89
Synonym Benzamide, 4-amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl]-4-oxido-2-morpholinyl]methyl]-, 4-Amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl]-4-oxido-2-morpholinyl]methyl]benzamide, 4-Amino-5-chloro-2-ethoxy-N-[[4-(4-fluorobenzyl)-4-oxido-2-morpholinyl]methyl]benzamide, Mosapride-4'-N-oxide, Mosapride N-oxide
IUPAC-namn 4-amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl]-4-oxidomorpholin-4-ium-2-yl]methyl]benzamide
LEDER CCOc1cc(N)c(Cl)cc1C(=O)NCC2C[N+]([O-])(Cc3ccc(F)cc3)CCO2
Specifikationer
Kemiskt namn eller material Mosapride N-oxide
Kvantitet 10 mg
InChI formel InChI=1S/C21H25ClFN3O4/c1-2-29-20-10-19(24)18(22)9-17(20)21(27)25-11-16-13-26(28,7-8-30-16)12-14-3-5-15(23)6-4-14/h3-6,9-10,16H,2,7-8,11-13,24H2,1H3,(H,25,27)
Formel vikt 437.1518