Läs mer
Thermo Scientific™ Motilin, Human, Porcine
Kvantitet:
0.5 mg
1 mg
Förpackningsstorlek:
0.5mg
1mg
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Art | Human, Porcine |
| Reningsmetod | HPLC |
| Innehåll och lagring | -30°C to -10°C |
| Formatera | Solid |
| Molekylvikt (g/mol) | 2714.10 |
| Produkttyp | Motilin, human, porcine |
| Kvantitet | 0.5 mg |
| Regulatorisk status | RUO - research use only |
| Sekvens | Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Met-Gln-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln |
For Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion