missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Motixafortide

Product Code. 16483986
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16483986 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483986 Supplier MedChemExpress Supplier No. HYP0171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Motixafortide (BKT140 4-fluorobenzoyl) is a novel CXCR4 antagonist with an IC50 vakue of ~1 nM.

Specifications

Renhet 98.27%
CAS 664334-36-5
Innehåll och lagring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
För användning med (applikation) Cancer-programmed cell death
Molekylvikt (g/mol) 2159.52 g/mol
Produkttyp Peptides
Kvantitet 25 mg
Frakt skick Room temperature
Löslighet DMSO : ≥ 100 mg/mL (46.31 mM) ∣H2O : 100 mg/mL (46.31 mM∣Need ultrasonic)
Synonym BKT140 (4-fluorobenzoyl)∣ BL-8040∣ TF14016
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.