Läs mer
R&D Systems™ Mouse ADAM2 (NP_033748.2) VersaClone cDNA
Specifikationer
Specifikationer
| Gen | ADAM2 |
| Tillträdesnummer | NP_033748.2 |
| Gen-ID (Entrez) | 2515 |
| Gene Alias | ADAM metallopeptidase domain 2, Cancer/testis antigen 15, CRYN2, CT15, CT15ADAM 2, disintegrin and metalloproteinase domain-containing protein 2, EC 3.4.24, fertilin beta, Fertilin subunit beta, FTNBCRYN1, PH30, PH-30, PH-30 b, PH30-beta, PH30PH30-beta |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Art | Mouse |
| Produktlinje | R&D Systems |
| Kvantitet | 10 μg |