missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse Angiopoietin-like Protein 4/ANGPTL4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB97971-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Mouse samples. It is validated for ELISA
Specifikationer
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| CHO-derived mouse Angiopoietin-like Protein 4/ANGPTL4 protein,, Gln24-Ser410, Accession # Q9Z1P8 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 547113 | |
| ELISA | |
| Q9Z1P8 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 51129 | |
| Mouse | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering