Learn More
Mouse anti-Bovine Ig (Light chain) Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA119384
1148.05 SEK valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Western Blot: Overnight at 4°C. Sample preparation: bovine serum, dilution 1:50 in Laemmli reducing buffer, boiled in water bath for 3 min. Reducing conditions, 12% separating gel. The antibody strongly reacts with bovine IgG light chains, weakly reacts with IgM.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Bovine Ig (Light chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Bovine, Ovine | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
IVA285-1 | |
Unconjugated | |
Iglc | |
Fraction of bovine immunoglobulin. | |
100 μg | |
Secondary | |
4° C, do not freeze | |
IgG1 |