Läs mer
EPCAM, Mouse, Monoclonal Antibody, Abnova™
Beskrivning
This gene encodes a carcinoma-associated antigen and is a member of a family that includes at least two type I membrane proteins. This antigen is expressed on most normal epithelial cells and gastrointestinal carcinomas and functions as a homotypic calcium-independent cell adhesion molecule. The antigen is being used as a target for immunotherapy treatment of human carcinomas. Mutations in this gene result in congenital tufting enteropathy. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | EPCAM |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Monoclonal |
| Konjugera | Unconjugated |
| Beskrivning | Mouse monoclonal antibody raised against partial recombinant protein of EPCAM. |
| Utspädning | ELISA (1:5000-1:20000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.4 (1% BSA, 50% glycerol, 0.02% sodium azide) |
| Gen | EPCAM |
| Gene Alias | 17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2 |
| Gensymboler | EPCAM |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.