Läs mer
GSK3B, Mouse, Monoclonal Antibody, Abnova™
Beskrivning
The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Specifikationer
Specifikationer
| Antigen | GSK3B |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Monoclonal |
| Konjugera | Unconjugated |
| Beskrivning | Mouse monoclonal antibody raised against partial recombinant protein of GSK3B. |
| Utspädning | ELISA (1:5000-1:20000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS, pH 7.4 (1% BSA, 50% glycerol, 0.02% sodium azide) |
| Gen | GSK3B |
| Gensymboler | GSK3B |
| Värd art | Mouse |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.